missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium isopropoxide, 99.9% (metals basis), Thermo Scientific™

Product Code. 11490760
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490760 1 g 1g
11400770 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490760 Supplier Thermo Scientific Alfa Aesar Supplier No. 044451.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21864-21-1
Molecular Formula LiNb(OC3H7)6
MDL Number MFCD00210626

Specifications

Boiling Point 140°C (0.2 mmHg)
Quantity 1 g
UN Number UN2925
Sensitivity Moisture sensitive
Formula Weight 454.38
Percent Purity 99.9%
Physical Form Solid
Assay (metals basis)
Chemical Name or Material Lithium niobium isopropoxide
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.