missing translation for 'onlineSavingsMsg'
Learn More

Lithium molybdenum oxide, 99+%

Product Code. 11366758
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366758 500 g 500g
11376758 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366758 Supplier Thermo Scientific Alfa Aesar Supplier No. 013427.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13568-40-6 | Li2MoO4 | 173.826 g/mol

Lithium molybdenum oxide is mainly used as an inhibitor in some types of industrial air conditioning. It is used in comparative property studies of battery systems. It has been used as a petroleum cracking catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13568-40-6
Molecular Formula Li2MoO4
Molecular Weight (g/mol) 173.826
MDL Number MFCD00016178
InChI Key NMHMDUCCVHOJQI-UHFFFAOYSA-N
Synonym lithium molybdate, dilithium 1+ ion molybdate, dilithium dioxido dioxo molybdenum, dilithotab molybdate, 2li.moo4
PubChem CID 6093689
IUPAC Name dilithium;dioxido(dioxo)molybdenum
SMILES [Li+].[Li+].[O-][Mo](=O)(=O)[O-]

Specifications

Melting Point 705°C
Color White
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 173.83
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.66 g/mL
Chemical Name or Material Lithium molybdenum oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.