missing translation for 'onlineSavingsMsg'
Learn More

Lithium methoxide, pure, 2.2M (10 wt%) solution in methanol, AcroSeal™

Product Code. 10283661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal Glass Bottle
AcroSeal Glass Bottle in Tin Can
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10283661 100 mL AcroSeal Glass Bottle in Tin Can 100mL
10611955 800 mL AcroSeal Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10283661 Supplier Thermo Scientific Acros Supplier No. 336751000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 865-34-9 | CH3LiO | 37.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-34-9 , 67-56-1
Molecular Formula CH3LiO
Molecular Weight (g/mol) 37.97
MDL Number MFCD00036357
InChI Key JILPJDVXYVTZDQ-UHFFFAOYSA-N
Synonym lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3
PubChem CID 4177230
IUPAC Name lithium;methanolate
SMILES [Li+].C[O-]

Specifications

Color Colorless
Linear Formula CH3OLi
Density 0.8500g/mL
Formula Weight 37.97
Percent Purity ≥10%
Grade Pure
Packaging AcroSeal Glass Bottle in Tin Can
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Lithium methoxide, 10% Solution in Methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.