missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium methoxide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
Description
Lithium methoxide is used in sol-gel synthesis of lithium borophosphates. It is also useful for deprotonation reactions in organic synthesis. Further, it serves as a mild base in organic synthesis especially in transesterifications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >500°C (decomposition) |
Color | White |
Linear Formula | LiOCH3 |
Quantity | 500 g |
UN Number | UN3206 |
Beilstein | 3552389 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in methanol. |
IUPAC Name | lithium;methanolate |
Formula Weight | 37.98 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'