missing translation for 'onlineSavingsMsg'
Learn More

Lithium Metaborate, Purum p.a., Anhydrous, Molar ratio (B2O3/Li2O): 0.95-1.05, Honeywell™

Product Code. 15646060 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15646060 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15646060 Supplier Honeywell Research Chemicals Supplier No. 62615100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 845°C
CAS 13453-69-5
Color White
Physical Form Powder
Quantity 100 g
Molecular Formula B4H14Li2O7
Linear Formula LiBO2
MDL Number MFCD00011089
UN Number NONH for all modes of transport
Synonym lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
InChI Key DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
IUPAC Name tetraboron heptahydrate dilithium
Molecular Weight (g/mol) 183.23
PubChem CID 123308
Formula Weight 49.75g/mol
Packaging Poly bottle
Chemical Name or Material Lithium metaborate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.