Learn More
Lithium metaborate, dried, 99%
Description
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.
Solubility
Soluble in water(27.5g/L).
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alkali metals, acid anhydrides, acids.
Specifications
Specifications
| Melting Point | 845°C |
| Quantity | 25 g |
| Linear Formula | LiBO2 |
| Solubility Information | Soluble in water(27.5g/L). |
| Formula Weight | 49.75 |
| Percent Purity | 99% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 1.397 |
| Chemical Name or Material | Lithium metaborate, dried |
Safety and Handling
- Lithium metaborate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Lithium metaborate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.