Learn More
Lithium metaborate, anhydrous, tech.
Description
Lithium Metaborate is commonly used as a fused compound or a component for an assay. It has been used for the analysis of wild rice for silicon, calcium, magnesium and potassium. Lithium metaborate was used to fuse with solid samples for decomposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 845°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Linear Formula | LiBO2 |
| Solubility Information | Soluble in water (25.7mg/ml at 20°C). |
| IUPAC Name | lithium;oxido(oxo)borane |
| Formula Weight | 49.75 |
| Grade | Technical |
| Odor | Odorless |
| Show More |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.