missing translation for 'onlineSavingsMsg'
Learn More

Lithium metaborate, ACS, 98.0-102.0%

Product Code. 11334277
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334277 250 g 250g
11344277 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334277 Supplier Thermo Scientific Alfa Aesar Supplier No. 036760.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol

It acts as a support for antioxidant property in human and animal tissues. Used in non-linear optic materials and cellular phones. It is used for measuring total elements in soil, geological, plant and other natural organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-69-5
Molecular Formula B4H14Li2O7
Molecular Weight (g/mol) 183.23
MDL Number MFCD00011089
InChI Key DPYKRXYVXDYLEY-UHFFFAOYSA-N
Synonym lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
PubChem CID 123308
SMILES [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

Specifications

Melting Point 845°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.0-102.0%
Linear Formula LiBO2
Solubility Information soluble in ethanol.
IUPAC Name lithium;oxido(oxo)borane
Formula Weight 49.75
Percent Purity 98.0 to 102.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.397 g/mL
Chemical Name or Material Lithium metaborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.