missing translation for 'onlineSavingsMsg'
Learn More

Lithium meta-Borate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15528004 Shop All J.T.Baker Products
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15528004

Brand: Avantor J.T.Baker 238205

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 13453-69-5
Quantity 100 g
Molecular Formula B4H14Li2O7
Synonym lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
InChI Key DPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
IUPAC Name lithium;oxido(oxo)borane
Molecular Weight (g/mol) 183.23
PubChem CID 123308
Formula Weight 49.75
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.40kg
Chemical Name or Material Lithium meta-Borate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.