missing translation for 'onlineSavingsMsg'
Learn More

Lithium Metaborate/Lithium Bromide (99.5%/0.50%) Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™

Product Code. 15236046 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Triple-leached LDPE bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15236046 1 kg Triple-leached LDPE bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15236046 Supplier SPEX Certiprep Supplier No. FFB000502

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Lithium Metaborate/Lithium Bromide (99.5%/0.50%), Pure Grade Fusion Flux for Sample Preparation, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Pure Grade Fusion Flux
  • 1 kg
  • Lithium Metaborate/Lithium Bromide (99.5%/0.50%)

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Quantity 1 kg
Grade Pure
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lithium Metaborate/Lithium Bromide, Fusion Flux Pure

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.