missing translation for 'onlineSavingsMsg'
Learn More

Lithium manganese(III,IV) oxide, 99.5% (metals basis)

Product Code. 11313768
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313768 100 g 100g
11323768 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313768 Supplier Thermo Scientific Alfa Aesar Supplier No. 040250.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12057-17-9 | LiMn2O4 | 180.81 g/mol

It is used as anode materials of high-performance lithium ion battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12057-17-9
Molecular Formula LiMn2O4
Molecular Weight (g/mol) 180.81
MDL Number MFCD01114233
InChI Key KEZPOJYZIWMPGY-UHFFFAOYSA-N

Specifications

Melting Point >400°C
Physical Form Powder
Quantity 100 g
Linear Formula LiMn2O4
Solubility Information Insoluble in water.
Formula Weight 180.81
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lithium manganese(III,IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.