missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Lithium-Ion Battery

Product Code. 16363677 Shop All Brand Products
missing translation for 'orderingAttributeHoverText'
:
Lithium-Ion Battery
This item is not returnable. View return policy

Product Code. 16363677

missing translation for 'mfr': BRANDTECH™ BRAND™ 705225

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with HandyStep™ Touch and HandyStep™ Touch S.

  • Accessory
  • For use with HandyStep™ Touch and Touch S Electronic Repeating Pipette
TRUSTED_SUSTAINABILITY

Specifications

Battery Type Lithium-ion
Description Replacement lithium-ion battery
Color Black/White
For Use With BRAND HandyStep Touch/Touch S
Material Plastic
Type Battery
For Use With (Equipment) HandyStep Touch / Touch S
Quantity 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.