missing translation for 'onlineSavingsMsg'
Learn More

Lithium Iodide, MP Biomedicals

Product Code. 11413462 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11413462 50 g 50g
11403462 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 11413462

Brand: MP Biomedicals 0215525950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Produces a hazy solution at 50% (w/v) in H2O

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
IUPAC Name lithium(1+) iodide
SMILES [Li+].[I-]

Specifications

Quantity 50 g
Formula Weight 133.8
Chemical Name or Material Lithium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.