missing translation for 'onlineSavingsMsg'
Learn More

Lithium iodide, anhydrous, 98+%, Thermo Scientific Chemicals

Catalog Number p-7051663
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
10377-51-2
ILi
133.84
MFCD00011092
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
66321
lithium;iodide
[Li+].[I-]
This item is not returnable. View return policy
10377-51-2
ILi
133.84
MFCD00011092
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
66321
lithium;iodide
[Li+].[I-]

As absorbent in refrigeration process, as a catalyst, and in photography

Lithium iodide is used as reagent in the cleavage of esters, ethers and epoxides, and carboxylation. It is used in scintillation counters. Lithium iodide is used as an electrolyte in high temperature batteries and artificial pacemakers. Phosphor, which is a complex of iodine, is used in neuron detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1170°C to 1190°C
Melting Point 449°C
Color White to Yellow
Physical Form Lump/Powder
Quantity 50 g
Linear Formula LiI
Merck Index 14,5535
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 133.84
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.955
Sensitivity Hygroscopic; Light sensitive
Density 4.076 g/mL
Chemical Name or Material Lithium iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'