missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Monohydrate, TRC

Product Code. 30012280
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012280 250 g 250g
30007274 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012280 Supplier Toronto Research Chemicals Supplier No. TRCL469125250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula HLiO.H2O
Molecular Weight (g/mol) 42
Synonym Lithium Hydroxide Hydrate;
IUPAC Name hydroxylithium;hydrate
SMILES O.[Li]O

Specifications

Quantity 250 g
InChI Formula InChI=1S/Li.2H2O/h;2*1H2/q+1;;/p-1
Formula Weight 42.0293
Chemical Name or Material Lithium Hydroxide Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.