missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Monohydrate, puriss p.a, ≥99.0% (T), Honeywell™ Fluka™

Product Code. 15696050 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly Bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15696050 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15696050 Supplier Honeywell-Fluka Supplier No. 62531250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 423°C
CAS 1310-66-3
Color Colorless
pH Alkaline
Physical Form Solid
Quantity 250 g
Molecular Formula H3LiO2
Linear Formula LiOH · H2O
MDL Number MFCD00149772
UN Number UN2680
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES [Li+].O.[OH-]
IUPAC Name lithium(1+) hydrate hydroxide
Molecular Weight (g/mol) 41.96
PubChem CID 168937
Formula Weight 41.96g/mol
Percent Purity ≥99.0% (T)
Odor Odorless
Packaging Poly Bottle
Density 1.80g/cm3
Chemical Name or Material Lithium hydroxide monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.