missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, LiOH 56.5% min

Product Code. p-4407570
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
This item is not returnable. View return policy

Product Code. 11336738

Brand: Thermo Scientific Alfa Aesar 013409.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Physical Form Powder
Quantity 50 g
Assay Percent Range 56.5% min
Solubility Information Soluble in water,methanol and ethanol.
IUPAC Name lithium(1+) hydrate hydroxide
Formula Weight 41.96 (23.95 Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Density 1.51 g/mL
Chemical Name or Material Lithium hydroxide monohydrate
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Lithium hydroxide monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.