missing translation for 'onlineSavingsMsg'
Learn More

Lithium Hydroxide Monohydrate, Solstice

Product Code. 15621240 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 1 kg
100 kg
Unit Size:
100g
100kg
1kg
500g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15621240 500 g 500g
15691230 100 g 100g
15611240 1 kg 1kg
15631240 6 x 1 kg Pack of 6
15601240 100 kg 100kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15621240 Supplier Honeywell-Fluka Supplier No. 13020500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum,≥98.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Quantity 500 g
Linear Formula LiOH · H2O
UN Number 2680
IUPAC Name lithium;hydroxide;hydrate
Formula Weight 41.96g/mol
Percent Purity ≥98.5%
Grade purum
Chemical Name or Material Lithium Hydroxide Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.