missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH

Product Code. 10582411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
25g
5kg
Product Code. Quantity Packaging unitSize
10582411 250 g Plastic bottle 250g
10226880 25 g Plastic bottle 25g
10378240 1 kg Plastic bottle 1kg
11964151 5 kg Plastic drum 5kg
4 options
This item is not returnable. View return policy

Product Code. 10582411

Brand: Thermo Scientific Acros 199542500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Boiling Point 920.0°C
Melting Point 462.0°C
Color Cream to White or Yellow
Physical Form Crystalline Powder
Quantity 250 g
Linear Formula LiOH·H2O
Fieser 07,208; 12,282
Merck Index 14,5534
Solubility Information Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol
IUPAC Name lithium(1+) hydrate hydroxide
Formula Weight 41.95
Percent Purity ∼56% (LiOH)
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium hydroxide monohydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.