missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, 98%

Product Code. 11462927
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462927 2500 g 2500g
11442927 100 g 100g
11452927 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462927 Supplier Thermo Scientific Alfa Aesar Supplier No. A15519.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

Solubility
Soluble in water, methanol and ethanol.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture, strong oxidizing agents, strong acids and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Specifications

Quantity 2500 g
UN Number UN2680
Merck Index 14,5534
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 41.96 (23.95 Anhydrous)
Percent Purity 98%
Sensitivity Air sensitive; Hygroscopic
Density 1.51
Chemical Name or Material Lithium hydroxide monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.