missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide monohydrate, LiOH 56.5% min

Product Code. 11346738
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346738 500 g 500g
11336738 50 g 50g
11326738 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346738 Supplier Thermo Scientific Alfa Aesar Supplier No. 013409.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Physical Form Powder
Quantity 500 g
Assay Percent Range 56.5% min
Solubility Information Soluble in water,methanol and ethanol.
IUPAC Name lithium;hydroxide;hydrate
Formula Weight 41.96 (23.95 Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Density 1.51 g/mL
Chemical Name or Material Lithium hydroxide monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.