missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, Honeywell™

Product Code. p-7072215 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15645820 100 g 100g
15655820 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15645820 Supplier Honeywell Research Chemicals Supplier No. 545856100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
IUPAC Name lithium(1+) hydroxide
SMILES [Li+].[OH-]

Specifications

Quantity 100 g
Linear Formula LiOH
UN Number UN2680
Formula Weight 23.95g/mol
Percent Purity ≥98%
Chemical Name or Material Lithium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.