missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, anhydrous, 98%

Product Code. 11396728
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396728 100 g 100g
15481037 25 g 25g
11386728 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396728 Supplier Thermo Scientific Alfa Aesar Supplier No. 013407.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-65-2 | HLiO | 23.95 g/mol

Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
SMILES [Li+].[OH-]

Specifications

Boiling Point 924°C
Melting Point 462°C
Color White
Quantity 100 g
Assay Percent Range 98%
Solubility Information Soluble in water. Slightly soluble in alcohol.
IUPAC Name lithium;hydroxide
Formula Weight 23.95
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance White powder
Density 2.54
Chemical Name or Material Lithium hydroxide, anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Lithium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.