missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, anhydrous, 98%, Thermo Scientific Chemicals

Catalog Number p-4407573
Click to view available options
Quantity:
25 g
100 g
500 g
1310-65-2
HLiO
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
3939
CHEBI:33979
[Li+].[OH-]
1310-65-2
HLiO
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
3939
CHEBI:33979
[Li+].[OH-]

Lithium hydroxide is used as a heat transfer medium, as a storage-battery electrolyte and also used for the production of lithium greases. It is also used in ceramics, in some portland cement formulations, in the absoption of carbondioxide from sealed enviornments such as submarines, spacecrafts and breathing apparatus. It is used in esterification reactions, as stabilizer in photographic developments and as a coolant in pressurized water reactors for corrosion control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 924°C
Melting Point 462°C
Color White
Quantity 25 g
Assay Percent Range 98%
Solubility Information Soluble in water. Slightly soluble in alcohol.
IUPAC Name lithium(1+) hydroxide
Formula Weight 23.95
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance White powder
Density 2.54
Chemical Name or Material Lithium hydroxide, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'