missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluorosilicate, Thermo Scientific Chemicals

Product Code. 11333748
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333748 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11333748 Supplier Thermo Scientific Alfa Aesar Supplier No. 039332.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lithium hexafluorosilicate is used as chemicals, pharmaceutical intermediates and reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17347-95-4
Molecular Formula F6LiSi
Molecular Weight (g/mol) 149.02
MDL Number MFCD00049473
InChI Key ZXUANORTLYQJEG-UHFFFAOYSA-N

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN2856
Solubility Information Soluble in water and methanol.
Formula Weight 155.96
Odor Odorless
Chemical Name or Material Lithium hexafluorosilicate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.