missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluoroarsenate(V), 99%

Product Code. 11357217
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357217 50 g 50g
11367217 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11357217 Supplier Thermo Scientific Alfa Aesar Supplier No. 011530.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 29935-35-1 | AsF6Li | 195.85 g/mol

Lithium hexafluoroarsenate(V) is a potential battery electrolyte material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29935-35-1
Molecular Formula AsF6Li
Molecular Weight (g/mol) 195.85
MDL Number MFCD00011076
InChI Key YYPUQABLGAAHDV-UHFFFAOYSA-H
Synonym lithium hexafluoroarsenate, lithium hexafluoroarsenate v, lithium hexafluoroarsoranuide, lithium hexakis fluoranyl arsenic 1-, lithotab hexafluoro-$l^ 5-arsanuide, lithium 1+ hexafluoro-??-arsanuide
PubChem CID 9837036
SMILES [Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]

Specifications

Physical Form Powder
Quantity 50 g
UN Number UN1557
Solubility Information Insoluble in water.
IUPAC Name λ⁵-arsenic(5+) lithium(1+) hexafluoride
Formula Weight 195.85
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material Lithium hexafluoroarsenate(V)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.