missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluoroantimonate, 97%, Thermo Scientific Chemicals

Product Code. 11358907
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358907 5 g 5g
11348907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358907 Supplier Thermo Scientific Alfa Aesar Supplier No. 038595.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lithium hexafluoroantimonate as a photo initiator for production of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18424-17-4
Molecular Formula F6LiSb
Molecular Weight (g/mol) 242.69
MDL Number MFCD00042319
InChI Key YVBBFYDVCPCHHG-UHFFFAOYSA-H
Synonym lithium hexafluoroantimonate, lithium 1+ ion hexafluorostibanuide, acmc-1bpt7, lithotab hexafluorostibanuide, lithium hexafluorostiboranuide, hexafluorostibine, lithium salt, lithium hexafluoroantimonate 1-, lithium hexakis fluoranyl antimony 1-
PubChem CID 16700586
IUPAC Name lithium;hexafluoroantimony(1-)
SMILES [Li+].F[Sb-](F)(F)(F)(F)F

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN1549
Solubility Information Very soluble in water
Formula Weight 242.68
Percent Purity 97%
Sensitivity Hygroscopic
Chemical Name or Material Lithium hexafluoroantimonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.