missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium hexafluoroantimonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Lithium hexafluoroantimonate as a photo initiator for production of polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | (decomposition) |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN1549 |
Solubility Information | Very soluble in water |
Formula Weight | 242.68 |
Percent Purity | 97% |
Sensitivity | Hygroscopic |
Chemical Name or Material | Lithium hexafluoroantimonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'