missing translation for 'onlineSavingsMsg'
Learn More

Lithium formate hydrate, 98%

Product Code. 11428017
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11428017

Brand: Thermo Scientific Alfa Aesar A17432.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6108-23-2 | CHLiO2 | 51.96 g/mol

It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the investigation of signal fading in lithium formate EPR dosimeters using a new sensitive method.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-23-2
Molecular Formula CHLiO2
Molecular Weight (g/mol) 51.96
MDL Number MFCD00050823
InChI Key XKPJKVVZOOEMPK-UHFFFAOYSA-M
Synonym lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
PubChem CID 23702287
IUPAC Name lithium;formate;hydrate
SMILES [Li+].[O-]C=O

Specifications

Melting Point 94°C (decomposition)
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 51.96 (Anhydrous)
Percent Purity 98%
Odor Mild
Chemical Name or Material Lithium formate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.