missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, Optical Grade, 99.5+%

Product Code. 11357667
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357667 5 g 5g
11367667 25 g 25g
11377667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357667 Supplier Thermo Scientific Alfa Aesar Supplier No. 014056.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-24-4 | FLi | 25.94 g/mol

Used in specialized UV optics. Also used as a crystal in X-ray spectrometry. Widely employed in PLED (Polymer Light Emitting Diodes) and OLED (Organic Light Emitting Diodes) as a coupling layer to enhance electron injection. Found in liquid-fluoride nuclear reactors. Used to record ionizing radiation in thermoluminescent dosimeters. Component of molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Physical Form Broken Crystals
Quantity 5 g
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in hydrofluoric acid. Less soluble in water. Insoluble in alcohol.
IUPAC Name lithium;fluoride
Formula Weight 25.94
Percent Purity ≥99.5%
Grade Optical
Odor Odorless
Refractive Index 1.3915
Density 2.635 g/mL
Chemical Name or Material Lithium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.