missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, Puratronic™, 99.99% (metals basis)

Product Code. 11481307
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
100g
10g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481307 10 g 100g
11376817 10 g 10g
11356817 50 g 50g
11346817 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11481307 Supplier Thermo Scientific Alfa Aesar Supplier No. B24260.22
This item is not returnable. View return policy

CAS: 7789-24-4 | FLi | 25.94 g/mol

Used in specialized UV optics. Also used as a crystal in X-ray spectrometry. Widely employed in PLED (Polymer Light Emitting Diodes) and OLED (Organic Light Emitting Diodes) as a coupling layer to enhance electron injection. Found in liquid-fluoride nuclear reactors. Used to record ionizing radiation in thermoluminescent dosimeters. Component of molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in HF. Less soluble in water. Insoluble in alcohol.
IUPAC Name lithium;fluoride
Formula Weight 25.94
Odor Odorless
Refractive Index 1.3915
Appearance White powder
Density 2.635 g/cm3 at 20°C
Chemical Name or Material Lithium fluoride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyethyl methacrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Hydroxyethyl methacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.