missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.85% (metals basis)

Product Code. 11303458
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303458 50 g 50g
11313458 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303458 Supplier Thermo Scientific Alfa Aesar Supplier No. 036359.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-24-4 | FLi | 25.94 g/mol

Used in the preparation of hypersensitive thermoluminescent materials, dosimeters and in the room-temperature oxidation of noble metals using fluorine in anhydrous HF. It is mainly used as a component of molten salts. Lithium fluoride forms the basic constituent of the preferred fluoride salt mixture used in liquid-fluoride nuclear reactors. Lithium fluoride is widely used in PLED and OLED as a coupling layer to enhance electron injection. LiF crystals are transparent to short wavelength ultraviolet radiation, more so than any other material. LiF is therefore used in specialized UV optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
IUPAC Name lithium;fluoride
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Lump/Powder
Quantity 50 g
Linear Formula LiF
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in 0.29g/100 mL (20°C) and hydrogen fluoride. Insoluble in alcohol.
Formula Weight 25.94
Percent Purity 99.85%
Odor Odorless
Refractive Index 1.3915
Density 2.635 g/mL
Assay (metals basis)
Chemical Name or Material Lithium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.