missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 97%

Product Code. 11434733
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11434733

Brand: Thermo Scientific Alfa Aesar A11632.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-24-4 | FLi | 25.94 g/mol

Lithium fluoride is widely used in many fields. It is used in the soldering progress for glass lining as a cosolvent. It finds application as an additive of aluminum electrolysis and rare earth electrolysis as a crystal in X-ray spectrometry. It is also used in specialized UV optics due to its large band gap and transparency to short wave length ultraviolet radiation. It is involved to record ionizing radiation exposure from gamma rays, beta particles and neutrons in thermoluminescent dosimeters. In nuclear reactors, lithium fluoride is mixed with beryllium fluoride to form a base solvent, which is used in molten-salt reactor experiment (MSRE). Additionally, it is useful as a coupling layer to enhance electron injection in polymer light-emitting diodes (PLED) and organic light-emitting diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium fluoride is widely used in many fields. It is used in the soldering progress for glass lining as a cosolvent. It finds application as an additive of aluminum electrolysis and rare earth electrolysis as a crystal in X-ray spectrometry. It is also used in specialized UV optics due to its large band gap and transparency to short wave length ultraviolet radiation. It is involved to record ionizing radiation exposure from gamma rays, beta particles and neutrons in thermoluminescent dosimeters. In nuclear reactors, lithium fluoride is mixed with beryllium fluoride to form a base solvent, which is used in molten-salt reactor experiment (MSRE). Additionally, it is useful as a coupling layer to enhance electron injection in polymer light-emitting diodes (PLED) and organic light-emitting diode (OLED).

Solubility
Soluble in water and hydrogen fluoride. Insoluble in alcohol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with aqueous solutions, strong acids and strong oxidizing agents. Hydrolyzes in the presence of water to form hydrofluoric acid which attacks glass.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
IUPAC Name lithium;fluoride
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Quantity 500 g
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in water and hydrogen fluoride. Insoluble in alcohol.
Formula Weight 25.94
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.3915
Density 2.635
Chemical Name or Material Lithium fluoride
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.