missing translation for 'onlineSavingsMsg'
Learn More

Lithium DL-2-aminoadipate, 97+%, Thermo Scientific™

Product Code. 11430081
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
Unit Size:
0.1g
0.5g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430081 0.5 g 0.5g
11450081 0.1 g 0.1g
11440081 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430081 Supplier Thermo Scientific Alfa Aesar Supplier No. 044797.02
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H9Li2NO4
MDL Number MFCD06657665
Synonym DL-2-Aminoadipic acid lithium salt

Specifications

Color White
Linear Formula [O2C(CH2)3CH(NH2)CO2]Li2
Quantity 0.5 g
Formula Weight 173.02
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Lithium DL-2-aminoadipate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.