missing translation for 'onlineSavingsMsg'
Learn More

Lithium dihydrogen phosphate, 97%, Thermo Scientific Chemicals

Product Code. 11458826
Click to view available options
Quantity:
1 kg
Unit Size:
1000g
This item is not returnable. View return policy

Product Code. 11458826

Brand: Thermo Scientific Alfa Aesar A16987.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store
TRUSTED_SUSTAINABILITY

Specifications

Melting Point >100°C
CAS 13453-80-0
Quantity 1 kg
Molecular Formula H2LiO4P
MDL Number MFCD00016188
Synonym lithium dihydrogen phosphate, monolithium phosphate, lithium dihydrophosphate, lithium dihydrogenorthophosphate, lithium dihydrogen orthophosphate, phosphoric acid, monolithium salt, lithium phosphate monobasic, phosphoric acid, lithium salt 1:1, lithiumdihydrogenphosphate, li.h2o4p
Solubility Information Slightly soluble aq. soln..
InChI Key SNKMVYBWZDHJHE-UHFFFAOYSA-M
SMILES [Li+].OP(O)([O-])=O
IUPAC Name lithium;dihydrogen phosphate
Molecular Weight (g/mol) 103.93
PubChem CID 23669251
Formula Weight 103.93
Percent Purity 97%
Odor Odorless
Density 2.461
Chemical Name or Material Lithium dihydrogen phosphate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.