missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Lithium cyclopentadienide is a useful reagent in preparing metallocenes and cyclopentadienyl derivatives. It is also used as intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 g |
UN Number | UN3180 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 72.04 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | Lithium cyclopentadienide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'