missing translation for 'onlineSavingsMsg'
Learn More

Lithium cobalt(III) oxide, 97%

Product Code. 11397657
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397657 1 kg 1kg
11387657 50 g 50g
11307667 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397657 Supplier Thermo Scientific Alfa Aesar Supplier No. 014049.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12190-79-3 | CoLiO2 | 97.871 g/mol

Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12190-79-3
Molecular Formula CoLiO2
Molecular Weight (g/mol) 97.871
MDL Number MFCD00049786
InChI Key BFZPBUKRYWOWDV-UHFFFAOYSA-N
Synonym lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
PubChem CID 23670860
IUPAC Name lithium;oxido(oxo)cobalt
SMILES [Li+].[O-][Co]=O

Specifications

Color Blue
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97%
Solubility Information Insoluble in water.
Formula Weight 97.88
Odor Odorless
Sensitivity Hygroscopic
Appearance Dark blue powder
Chemical Name or Material Lithium cobalt(III) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.