missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, ultra dry, 99.9% (metals basis)

Product Code. 11360058
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360058 100 g 100g
11350058 5 g 5g
11370058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11360058 Supplier Thermo Scientific Alfa Aesar Supplier No. 042842.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7447-41-8 | ClLi | 42.39 g/mol

Lithium chloride is useful for the production of lithium metal, and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer, and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere, and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis, and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1325°C to 1360°C
Melting Point 605°C
Color White to Gray
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Freely soluble in water; soluble in methanol,ethanol,formic acid. Insoluble in dichloromethane.
Formula Weight 42.39
Odor Odorless
Sensitivity Hygroscopic
Appearance White to gray crystalline powder
Density 2.068
Chemical Name or Material Lithium chloride, ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.