missing translation for 'onlineSavingsMsg'
Learn More

Electrode Filling Solution, Lithium Chloride in Glacial Acetic Acid, Reagecon™

Used for pH measurement or titration in non-aqueous media or organic solvents. Electrode Filling Solution, Lithium Chloride in Glacial Acetic Acid, Reagecon™ is a 1M solution of LiCl in CH3COOH for filling the inner and outer chamber of refillable pH electrodes.

Brand:  Reagecon EFSLIGA

Code : 04

Additional Details : CAS Number : 64-19-7 Weight : 0.10000kg Transport : UN number : 2789 Chem class : 8 Pack group : II


Product Code. 11573172

  • £27.35 / Each
Estimated Shipment: 22-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lithium chloride is an ionic salt that forms a non-aqueous electrolyte solution. While potassium chloride solution is a more typical electrode filling solution, use of lithium chloride in glacial acetic acid is indicated when the electrode is to be used in non-aqueous media and organic solvents.
Specifications

Specifications

Lithium chloride
pH Electrode
100 mL
1M Lithium chloride (LiCl) dissolved 100ml in glacial acetic acid
42.39g/mol
7447-41-8,64-19-7
Liquid
Electrode Filling
LiCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ELECTRODE FILLING SOLUTION 1.0M LITHIUM CHLORIDE IN GLACIAL ACETIC ACID

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : acetic acid, lithiumchloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers