missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)

Product Code. 11336817
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336817 25 g 25g
11326817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336817 Supplier Thermo Scientific Alfa Aesar Supplier No. 010735.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85144-11-2 | ClLi | 42.39 g/mol

Lithium chloride is widely employed in lithium metal production. It is used as a brazing flux for aluminum in automobile parts. Its use can also be seen as a desiccant for drying air streams. In biochemical applications, lithium chloride is used to precipitate RNA from cellular extracts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85144-11-2
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00149764
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
PubChem CID 23681138
SMILES [Li+].[Cl-]

Specifications

Melting Point >98°C-H2O
Color White
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water.
IUPAC Name lithium;chloride;hydrate
Formula Weight 42.39 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Lithium chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.