missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)

Product Code. 11326817
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11326817

Brand: Thermo Scientific Alfa Aesar 010735.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85144-11-2 | ClLi | 42.39 g/mol

Lithium chloride is widely employed in lithium metal production. It is used as a brazing flux for aluminum in automobile parts. Its use can also be seen as a desiccant for drying air streams. In biochemical applications, lithium chloride is used to precipitate RNA from cellular extracts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85144-11-2
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00149764
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
PubChem CID 23681138
SMILES [Li+].[Cl-]

Specifications

Melting Point >98°C-H2O
Color White
Physical Form Crystalline Aggregates
Quantity 100 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water.
IUPAC Name lithium(1+) chloride
Formula Weight 42.39 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Lithium chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.