missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, ACS Reagent, Honeywell Fluka™

Catalog Number 15693180
Click to view available options
Quantity:
5 g
2.5 kg
This item is not returnable. View return policy
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
This item is not returnable. View return policy
7447-41-8
ClLi
42.39
MFCD00011078
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 2.5 kg
Linear Formula LiCl
UN Number NONH for all modes of transport
Formula Weight 42.39g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Lithium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'