missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, ACS reagent, ≥99%, Solstice

Product Code. 16655793
Change view
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16655793 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16655793 Supplier Fluka Supplier No. 31046810KG
  • ONLINE EXCLUSIVE PRICE
    £3030.00
    £2789.53 / 10kg
    Save £240.47
    7% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 1,360°C (1,013 hPa)
Melting Point 605°C
CAS 7447-41-8
Color Colorless
CAS Max % ≤ 100%
pH 6 to 8
Quantity 10 kg
Molecular Formula ClLi
Linear Formula LiCl
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES [Li+].[Cl-]
IUPAC Name lithium(1+) chloride
Molecular Weight (g/mol) 42.39
Grade ACS
Density 2.07 g/cm3 (20°C)
Chemical Name or Material Lithium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.