missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, ≥99%, MP Biomedicals

Product Code. 11416481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416481 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11416481 Supplier MP Biomedicals Supplier No. 0219401090
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity lithium chloride reagent for general laboratory use.

  • Assay: ≥99%
  • Molecular Formula: ClLi
  • Formula Weight: 42.39 g/mol
  • CAS Number: 7447-41-8

Specifications

Boiling Point 2417 to 2480°F
CAS 7447-41-8
Color White
Physical Form Crystalline Powder
Quantity 500 g
Packaging Bottle
Appearance White Crystalline Powder
Assay Percent Range ≥99%
Decomposition Information Hydrogen Chloride, Lithium Oxide
Molecular Formula ClLi
Linear Formula LiCl
MDL Number MFCD00011078
UN Number NONH for all modes of transport
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
Solubility Information Soluble in water
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES [Li+].[Cl-]
IUPAC Name lithium(1+) chloride
Molecular Weight (g/mol) 42.39
PubChem CID 433294
ChEBI CHEBI:48607
Formula Weight 42.4
Percent Purity ≥99%
Chemical Name or Material Lithium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.