missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals

Product Code. 10532031
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

Lithium Chloride, 99%, ClLi, CAS Number-7447-41-8, chlorure de lithium, chlorku litu polish, chlorku litu, lithium chloride licl, lithium chloride, licl, luthium chloride, lithiumchloride, lithiumchlorid, chlorolithium, 500g, carbonate (as Li2CO3): < 0.03% (typically), 98.5% min. (Argentometry)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382.0°C
Melting Point 605.0°C
Color White
Physical Form Crystalline Granules
Quantity 100 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15,5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
Formula Weight 42.38
Percent Purity 99%
Grade Analytical
Loss on Drying 0.5% max. (400°C, 4 hrs)
Packaging Plastic bottle
Specific Gravity 2.06
Density 2.0600g/mL
Chemical Name or Material Lithium chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'