missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium chloride, 99%, For Analysis, Anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1382.0°C |
Melting Point | 605.0°C |
Color | White |
Physical Form | Crystalline Granule |
Quantity | 500 g |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | LiCl |
Fieser | 01,609; 02,246; 04,298; 05,677; 12,277; 16,194 |
Merck Index | 15,5584 |
Solubility Information | Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'