missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, ≥99.0% (AT), Honeywell™ Fluka™

Product Code. 15666920 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Poly bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 15666920

Brand: Honeywell-Fluka 730365G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., anhydrous,≥99.0% (AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1360°C
Melting Point 605°C
Color Colorless
pH 6.0 to 8.0
Physical Form Solid
Quantity 5 g
Linear Formula LiCl
UN Number NONH for all modes of transport
Solubility Information Soluble
IUPAC Name lithium;chloride
Formula Weight 42.39g/mol
Percent Purity ≥99.0% (AT)
Odor Odorless
Packaging Glass bottle
Density 2.07g/cm3
Chemical Name or Material Lithium chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.