missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, 200mM aq. soln., Thermo Scientific™

Product Code. 15484969
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 15484969

Brand: Thermo Scientific Alfa Aesar J62644.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in formation of an active Mn(0) species useful for radical cyclization reactions. It is utilized in a variety of assays to study cell-fate and neurobiology. In developing Xenopus embryos, lithium chloride is observed to exert inhibition of GSK-3β (glycogen synthase kinase-3β). Additionally, in transfected Chinese Hamster Ovary cells (CHO) stimulated with angiotension II (Ang II), it was noted that lithium chloride enhanced the production of inositol triphosphate. The antiviral properties of lithium chloride were noted in a study which showed that the compound inhibited pseudorabis virus infection in vitro. It is primarily used in the synthesis of lithium metal by electrolysis. Also acts as a prescribed drug for bipolar mood disorder, it may also exhibit anti-inflammatory properties.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, pyridine, nitrobenzene, acetone, and amyl alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
Linear Formula LiCl
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol,ether,pyridine,nitrobenzene,acetone,and amyl alcohol.
IUPAC Name lithium(1+) chloride
Formula Weight 42.39
Concentration or Composition (by Analyte or Components) 200mM aq. soln.
Odor Odorless
Chemical Name or Material Lithium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.