missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Carbonate, purum p.a, ≥98.0% (T), Honeywell™
Purum p.a.,≥98.0% (T)
£78.99 - £163.91
Chemical Identifiers
CAS | 554-13-2 |
---|---|
Molecular Formula | CLi2O3 |
Molecular Weight (g/mol) | 73.89 |
MDL Number | MFCD00011084 |
InChI Key | XGZVUEUWXADBQD-UHFFFAOYSA-L |
Synonym | lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine |
PubChem CID | 11125 |
ChEBI | CHEBI:6504 |
IUPAC Name | dilithium;carbonate |
SMILES | [Li+].[Li+].[O-]C([O-])=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
554-13-2 | |
73.89 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
11125 | |
dilithium;carbonate |
CLi2O3 | |
MFCD00011084 | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
CHEBI:6504 | |
[Li+].[Li+].[O-]C([O-])=O |
Specifications
1300°C | |
554-13-2 | |
9 to 11 (at 1g/L, 20°C) | |
CLi2O3 | |
MFCD00011084 | |
3999191 | |
XGZVUEUWXADBQD-UHFFFAOYSA-L | |
dilithium;carbonate | |
11125 | |
73.89g/mol | |
Odorless | |
Lithium carbonate |
720°C | |
White | |
Crystalline | |
Li2CO3 | |
NONH for all modes of transport | |
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine | |
[Li+].[Li+].[O-]C([O-])=O | |
73.89 | |
CHEBI:6504 | |
≥98.0% (T) | |
2.11g/cm3 |
Safety and Handling
P305 + P351 + P338
missing translation for 'rtecsNumber' : 209-062-5