missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, MP Biomedicals

Product Code. 11412216 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412216 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412216 Supplier MP Biomedicals Supplier No. 0521261280
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity lithium carbonate for research and laboratory use.

  • CAS Number: 554-13-2
  • Molecular Formula: CLi2O3
  • Molecular Weight: 73.888 g/mol
  • Melting Point: 723°C
  • Boiling Point: Decomposes at 1,300°C
  • Density: 2.11 g/cu cm
  • Solubility in water: 1.52 wt% at 0°C
  • Usage Statement: For research or further manufacturing use only, not for direct human use.

Tekniske data

Melting Point 723°C
CAS 554-13-2
Quantity 100 g
Odor Odorless
Specific Gravity 2.11
Density 2.11 g/cm³
Molecular Formula CLi2O3
Linear Formula Li2CO3
MDL Number MFCD00011084
Beilstein 3999191
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
Solubility Information Soluble in water
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES [Li+].[Li+].[O-]C([O-])=O
IUPAC Name dilithium(1+) carbonate
Molecular Weight (g/mol) 73.89
PubChem CID 11125
ChEBI CHEBI:6504
Formula Weight 73.89
Chemical Name or Material Lithium Carbonate
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.